ContactUsLinkComponent
In addition to 5'-3' DNA polymerase activity, it also possesses 3'-5' exonuclease activity. pfu DNA polymerase exhibits the lowest error rate of any thermostable DNA polymerase studied, is even up to ten fold more accurate than normal taq DNA polymer 1 * 1.000 EU
Catalog # ABNOD0014-2
Supplier:  Abnova
In addition to 5'-3' DNA polymerase activity, it also possesses 3'-5' exonuclease activity. pfu DNA polymerase exhibits the lowest error rate of any thermostable DNA polymerase studied, is even up to ten fold more accurate than normal taq DNA polymer 1 * 1.000 EU
Catalog # ABNOD0014-2
Supplier:  Abnova
Frequently Bought Together