LONGTAQ DNA POLYMERASE CATALYZES 5'-3' SYNTHESIS OF DNA. THE ENZYME HAS BEEN PROVED NOT HAVING THE 3'-5' EXONUCLEASE ACTIVITY. THE E NZYME HAS PROVEN TO HAVE HIGH AMPLIFICATION YIELD, BE STABLE AT HIGH TEMPARETAURE. 1 * 1.000 EU
ABNOD0013-2
: Abnova
Frequently Bought Together