TAQ DNA POLYMERASE CATALYZES 5'-3' SYNTHESIS OF DNA. THE ENZYME HAS BEEN PROVED NOT HAVING THE 3'-5' EXONUCLEASE ACTIVITY. THE ENZYM E HAS PROVEN TO HAVE HIGH AMPLIFICATION YIELD, BE STABLE AT HIGH TEMPARETAURE. 1 * 1.000 EU
Catalog # ABNOD0010-2
Supplier: Abnova
Frequently Bought Together