Digital Solutions
Applications
Support
About
Slovenia
Orders Link
ContactUsLinkComponent
Pipette Tip Boxes and Racks
Boxes for pipette tips
Boxes for pipette tips
Empty TipBox, PP, with tip-tray, stackable, for tips up to 50 µl, Grey
613-2424
:
Brand
Boxes for pipette tips
613-2424
:
Brand
Boxes for pipette tips
Empty TipBox, PP, with tip-tray, stackable, for tips up to 50 µl, Grey
4,80 €
1 * 1 items
-
+
Brand Pipette Tips and Filter Tips
Colour code:
Grey
Description:
Empty TipBox, PP, with tip-tray, stackable, for tips up to 50 µl
Seq:
01
Pipette Tips, Standard
#: 613-2429
: Brand