Orders LinkContactUsLinkComponent
Pipette tips
Pipette tips
Eppendorf type, macro, with collar, 1000 - 5000 µl, -, Transparent, Biohit, Dsg, Eppendorf, Genex, Jencons, Socorex Acura 835, 4 racks, 50 each
Catalog # DELT199075
Supplier:   DELTALAB
Pipette tips
Catalog # DELT199075
Supplier:  DELTALAB
Pipette tips
Eppendorf type, macro, with collar, 1000 - 5000 µl, -, Transparent, Biohit, Dsg, Eppendorf, Genex, Jencons, Socorex Acura 835, 4 racks, 50 each
Price
139,00 €
Log in for availability
1 * 4 items
Product Details & Documents
Specifications
  • Packed:
    4 racks, 50 each
  • Sterile:
    -
  • Volume:
    1000 - 5000 µl
  • Tip style:
    Eppendorf type, macro, with collar
  • Tip colour:
    Transparent
  • For use with:
    Biohit, Dsg, Eppendorf, Genex, Jencons, Socorex Acura 835