Biotix 200ul pipettetips. stack rack, pre-sterile, low retention, 5 layers of 192 tips/pack, 5 packs/case. universal fit ideal for both single and multichannel pipettors, graduation marks, patented features: Flexfit™ provides universal fit and super 1 * 960 items
Catalog # 613-2439
Supplier: BIOTIX
Frequently Bought Together