Orders LinkContactUsLinkComponent
BD Eclipse™ Needles
BD Eclipse™ Needles
Needle, sterile, 100/box, 1200/case, 21 G, 40 mm
Catalog # BDAM305895
Supplier:   BD Medical
BD Eclipse™ Needles
Catalog # BDAM305895
Supplier:  BD Medical
BD Eclipse™ Needles
BD Eclipse™ Needles
Needle, sterile, 100/box, 1200/case, 21 G, 40 mm
Price
518,00 €
Log in for availability
1 * 1.200 items
Product Details & Documents
The BD Eclipse™ SmartSlip™ is a single use sterile needle with a safety mechanism that covers the needle point after use. In the activated position, the needle cover guards against accidental needlestick during normal handling and disposal of the used needle/syringe.
  • The SmartSlip™ Technology is specially designed for safe assembling of BD Eclipse™ needles to Luer slip syringes
  • Bevel oriented to the safety cover to allow low angle injections
  • By assembling BD Eclipse™ needles to a luer slip syringe, an audible click indicates a safely assembled needle
  • Safety can be activated immediately after injection
  • Wide, textured finger pad for thumb or forefinger
  • Single-handed with one finger activation of the safety shield, audible click indicates that the cover has safely engaged the needle
  • Natural Rubber Latex is not part of the material formulation
  • Sterile, single use
The SmartSlip™ Technology consists of an adaptor to enable connection of the BD Eclipse safety needle to a Luer slip.
Specifications
  • Gauge:
    21 G
  • Description:
    Needle, sterile, 100/box, 1200/case
  • Length:
    40 mm
  • Seq:
    09