Orders LinkContactUsLinkComponent
VWR® Media Supplements for Dehydrated Culture Media
VWR® Media Supplements for Dehydrated Culture Media
CFC selective supplement, Pseudomonas agar base Cat. No. 84650.0500 - in compliance with ISO 13720 (meat products), 10 Vial
Catalog # 84746.0001
Supplier:   VWR Chemicals
VWR® Media Supplements for Dehydrated Culture Media
Catalog # 84746.0001
Supplier:  VWR Chemicals
CAS Number:  
VWR® Media Supplements for Dehydrated Culture Media
VWR® Media Supplements for Dehydrated Culture Media
CFC selective supplement, Pseudomonas agar base Cat. No. 84650.0500 - in compliance with ISO 13720 (meat products), 10 Vial
CAS Number:  
Price
82,80 €
Log in for availability
1 * 10 Vial
Product Details & Documents
The isolation of microorganisms often requires the use of specific supplements. The supplements presented here are available in vials and bottles, and the corresponding culture medium to use with them is indicated in the table below. Usually each vial is used to supplement 500 ml of media unless otherwise stated (complete information available on the specification sheets available on request).
  • Freeze-dried
Specifications
Specifications
  • Pk:
    10 Vial
  • Description:
    CFC selective supplement
  • For:
    Pseudomonas agar base Cat. No. 84650.0500 - in compliance with ISO 13720 (meat products)