VWR® Media Supplements for Dehydrated Culture Media
CFC selective supplement, Pseudomonas agar base Cat. No. 84650.0500 - in compliance with ISO 13720 (meat products), 10 Vial
Supplier: VWR Chemicals
VWR® Media Supplements for Dehydrated Culture Media
Catalog # 84746.0001
Supplier: VWR Chemicals
CAS Number:
Jump to:
Product Details & Documents
The isolation of microorganisms often requires the use of specific supplements. The supplements presented here are available in vials and bottles, and the corresponding culture medium to use with them is indicated in the table below. Usually each vial is used to supplement 500 ml of media unless otherwise stated (complete information available on the specification sheets available on request).
- Freeze-dried
Documents
Certificates
Search by Lot Number23071768.pdf SDS_SL_SI_VWRC301950.pdf SDS_SL_SI_VWRC84720 SDS_SL_SI_VWRC84720.pdf SDS_SL_SI_VWRC84721
SDS
More
301950ZK_TTC STERILE SOLUTION 1%_ISO 11133_rev1 84720.0001_FRASER SELECTIVE SUPPLEMENT_ISO 11133_rev1 84721.0001_FRASER HALF SELECTIVE SUPPLEMENT_ISO 11133_rev1 84722.0001_NOVOBIOCIN MKTTN SUPPLEMENT_ISO 11133_rev1 84723.0001_IODINE IODIDE SOLUTION_ISO 11133_rev1
More
spec Avantor 84726.0001 Spec VWRC84742.0001.pdf spec_84720.0001 spec_84721.0001 spec_84722.0001
Product specification
More
Specifications
Specifications
- Pk:10 Vial
- Description:CFC selective supplement
- For:Pseudomonas agar base Cat. No. 84650.0500 - in compliance with ISO 13720 (meat products)
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.