ContactUsLinkComponent
Waters reagent, RapiGest™ SF, drastically improves in-solution protein digestion in terms of speed and peptide recovery. RapiGest™ SF is available in 1mg vials containing enough material for 1-10 digestions. Add RapiGest™ SF to your proteolytic diges 1 * 10 mg
Catalog # WATE186002123
Supplier:  WATERS
Waters reagent, RapiGest™ SF, drastically improves in-solution protein digestion in terms of speed and peptide recovery. RapiGest™ SF is available in 1mg vials containing enough material for 1-10 digestions. Add RapiGest™ SF to your proteolytic diges 1 * 10 mg
Catalog # WATE186002123
Supplier:  WATERS
Frequently Bought Together