ContactUsLinkComponent
Solid, one piece molded foam, (eva) ice pan. Unbreakable containment for variety of temperature media such as ice, dry ice, liquid nitrogen, saline solution, and acetone. 1 L capacity, lime green. 1 * 1 items
Catalog # BCISBCS-211GR
Supplier:  BIOCISION
Solid, one piece molded foam, (eva) ice pan. Unbreakable containment for variety of temperature media such as ice, dry ice, liquid nitrogen, saline solution, and acetone. 1 L capacity, lime green. 1 * 1 items
Catalog # BCISBCS-211GR
Supplier:  BIOCISION
Frequently Bought Together