Mitosis and Meiosis Set II, - 5 selected Microscope SlidesMitosis, l.s. from Vicia faba (bean) root tips showing all mitotic stages. Iron hematoxyline stained - Lilium, anther t.s., microspore mother cells showing telophase of first and prophase of p 1 * 1 items
LIED5170
Frequently Bought Together