Orders LinkContactUsLinkComponent
Measuring scoops
Measuring scoops
Set of 8 coloured scoops (white, red, grey, black, yellow, dark blue, light blue, green), 100 ml, Assorted
Catalog # 231-0562
Supplier:   VITLAB
Measuring scoops
Catalog # 231-0562
Supplier:  VITLAB
Measuring scoops
Measuring scoops
Set of 8 coloured scoops (white, red, grey, black, yellow, dark blue, light blue, green), 100 ml, Assorted
Price
15,40 €
Log in for availability
1 * 1 items
Product Details & Documents
PP, coloured. With precision formed filling edge and comfortable, stable handle.
  • Also suitable as weighing scoops
  • Easily readable volume quantities visible on the upper side of the handle
  • Food-safe product according to EC Directive No. 10/2011
Specifications
  • Capacity:
    100 ml
  • Colour:
    Assorted
  • Description:
    Set of 8 coloured scoops (white, red, grey, black, yellow, dark blue, light blue, green)