ContactUsLinkComponent
recombinant/native protein, bub1b (human) recombinant protein, human bub1b (np_001202.4, 1 a.a. - 1050 a.a.) full-length recombinant protein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
  ABNOP5502
 :  Abnova
recombinant/native protein, bub1b (human) recombinant protein, human bub1b (np_001202.4, 1 a.a. - 1050 a.a.) full-length recombinant protein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
  ABNOP5502
 :  Abnova
Frequently Bought Together