ContactUsLinkComponent
recombinant protein, pdcd1 (human) recombinant protein, purified pdcd1 (aah74740.1 22 a.a. - 170 a.a.) human recombinant protein wit h his-flag-strepii tag at n-terminus expressed in human cells. 1 * 25 µG
Catalog # ABNOH00005133-H02
Supplier:  Abnova
recombinant protein, pdcd1 (human) recombinant protein, purified pdcd1 (aah74740.1 22 a.a. - 170 a.a.) human recombinant protein wit h his-flag-strepii tag at n-terminus expressed in human cells. 1 * 25 µG
Catalog # ABNOH00005133-H02
Supplier:  Abnova
Related Information
Frequently Bought Together