Orders LinkContactUsLinkComponent
Microscope slides, SuperFrost®, Menzel Gläser
Microscope slides, SuperFrost®, Menzel Gläser
Blue, Cellophane wrapped slides
Catalog # 630-2835
Supplier:   Epredia
Microscope slides, SuperFrost®, Menzel Gläser
Catalog # 630-2835
Supplier:  Epredia
Microscope slides, SuperFrost®, Menzel Gläser
Microscope slides, SuperFrost®, Menzel Gläser
Blue, Cellophane wrapped slides
Price
5,80 €
Log in for availability
1 * 50 items
Product Details & Documents
These slides are made of extra-white glass, washed clean and polished.
  • Labelling area resistant to all common solvents
  • Six colours for identification and colour coding
  • Thick labelling area to prevent slides from sticking together or getting scratched
Suitable for in vitro diagnostic (IVD) applications.
Complies with ISO 8037/I
Certifications: ISO 8037/I.
Packaging: In cardboard boxes of 50.
Specifications
  • Packed:
    Cellophane wrapped slides
  • Colour:
    Blue
  • Seq:
    11