Orders LinkContactUsLinkComponent
Assortment Boxes, PP
Assortment Boxes, PP
With 24 compartments, Blue, 335×225×55 mm, PP
Catalog # HUEN611900
Supplier:   hünersdorff GmbH
Assortment Boxes, PP
Catalog # HUEN611900
Supplier:  hünersdorff GmbH
Assortment Boxes, PP
Assortment Boxes, PP
With 24 compartments, Blue, 335×225×55 mm, PP
Price
18,30 €
Log in for availability
1 * 1 items
Product Details & Documents
Assortment box with carrying handle made of tough polypropylene.
  • Coloured bottom made of impact-proof, fracture-proof polypropylene
  • Food-safe colours, use in practically all areas of life possible
  • Resistant to cold and customary acids and alkalis
  • Highly transparent, tightly closing lid avoids mixing of the contents
Specifications
  • Colour:
    Blue
  • Description:
    With 24 compartments
  • Material:
    PP
  • Seq:
    02
  • W×D×H:
    335×225×55 mm