Orders LinkContactUsLinkComponent
VWR®, Tip Boxes, Empty
VWR®, Tip Boxes, Empty
Tip-Box N, PP, with push on lid, stackable and autoclavable, for 1000 µl tips, 50 - 1000 µl tips
Catalog # 613-2288
Supplier:   VWR Collection
VWR®, Tip Boxes, Empty
Catalog # 613-2288
Supplier:  VWR Collection
VWR®, Tip Boxes, Empty
VWR®, Tip Boxes, Empty
Tip-Box N, PP, with push on lid, stackable and autoclavable, for 1000 µl tips, 50 - 1000 µl tips
Price
15,50 €
Log in for availability
1 * 1 items
Product Details & Documents
Documents
Specifications
  • Description:
    Tip-Box N, PP, with push on lid, stackable and autoclavable, for 1000 µl tips
  • Seq:
    02
  • For use with:
    50 - 1000 µl tips