Process documentaion
APH 301 Connection cable type 1 for connection of a machine with serial interface to a PC. Crossover cable, length 3 m, Ø 2,5×80 mm, All glassware washers
Catalog # 141-0618
Supplier: MIELE
Jump to:
Product Details & Documents
Specifications
- Description:APH 301 Connection cable type 1 for connection of a machine with serial interface to a PC. Crossover cable, length 3 m, Ø 2,5×80 mm
- Seq:14
- For:All glassware washers
- Cat. No.:141-0618
- W×D×H:Ø 2.5×80 mm