Digital Solutions
Applications
Support
About
Slovenia
Orders Link
ContactUsLinkComponent
Glassware Washing Machines
Inserts
Inserts
Inserts
E 109 Insert suitable for 21 glass beakers up to 250 ml, equipped with 21×3 stay bars
141-0297
:
MIELE
Inserts
141-0297
:
MIELE
Expand image
10+
Inserts
E 109 Insert suitable for 21 glass beakers up to 250 ml, equipped with 21×3 stay bars
Add to cart
296,00 €
Unit of Measure
1 * 1 items
-
+
Add to cart
Description:
E 109 Insert suitable for 21 glass beakers up to 250 ml, equipped with 21×3 stay bars
Seq:
35