Orders LinkContactUsLinkComponent
Inserts
Inserts
E 963 insert, with 33×3 holders for beakers up to 250 ml, G 7827, 357×522×155 mm
Catalog # 141-0471
Supplier:   MIELE
Inserts
Catalog # 141-0471
Supplier:  MIELE
Inserts
Inserts
E 963 insert, with 33×3 holders for beakers up to 250 ml, G 7827, 357×522×155 mm
Price
1 224,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Description:
    E 963 insert, with 33×3 holders for beakers up to 250 ml
  • Seq:
    22
  • W×D×H:
    357×522×155 mm
  • For:
    G 7827