Orders LinkContactUsLinkComponent
Inserts
Insert E 960/1, with 20 large and 26 small spring hook for wide-necked Erlenmeyer flask and measuring cylinder, G 7827, 357×522×185 mm
  141-0479
 :   MIELE
Inserts
  141-0479
 :  MIELE
Inserts
Inserts
Insert E 960/1, with 20 large and 26 small spring hook for wide-necked Erlenmeyer flask and measuring cylinder, G 7827, 357×522×185 mm
1 661,00 €
1 * 1 items
  • Description:
    Insert E 960/1, with 20 large and 26 small spring hook for wide-necked Erlenmeyer flask and measuring cylinder
  • Seq:
    46
  • W×D×H:
    357×522×185 mm
  • For:
    G 7827