ContactUsLinkComponent
Cell Biology grade DMSO in dual labeled bottle to prepare concentrations from 5-20%. Use for preparation of cryopreservation media sterile, endotoxin-free. 1 * 1 items
  BCISBCS-3010
 :  BIOCISION
Cell Biology grade DMSO in dual labeled bottle to prepare concentrations from 5-20%. Use for preparation of cryopreservation media sterile, endotoxin-free. 1 * 1 items
  BCISBCS-3010
 :  BIOCISION
Frequently Bought Together