Pipette tips
Eppendorf type, 5 - 200 µl, -, Yellow, Eppendorf, Gilson, Socorex, Nichiryo, Biohit, Brand, Bulk (25×1000)
Supplier: DELTALAB
Jump to:
Product Details & Documents
Documents
Specifications
- Packed:Bulk (25×1000)
- Sterile:-
- Volume:5 - 200 µl
- Seq:09
- Tip style:Eppendorf type
- Tip colour:Yellow
- For use with:Eppendorf, Gilson, Socorex, Nichiryo, Biohit, Brand