Orders LinkContactUsLinkComponent
Pipette tips
Eppendorf type, crystal tip, bevelled, 0,5 - 10 µl, -, Transparent, Gilson, Eppendorf, Socorex, Nichiryo, 10 racks, 96 each
Catalog # DELT301-10
Supplier:   DELTALAB
Pipette tips
Catalog # DELT301-10
Supplier:  DELTALAB
Pipette tips
Eppendorf type, crystal tip, bevelled, 0,5 - 10 µl, -, Transparent, Gilson, Eppendorf, Socorex, Nichiryo, 10 racks, 96 each
Price
167,00 €
Log in for availability
1 * 10 items
Specifications
  • Packed:
    10 racks, 96 each
  • Sterile:
    -
  • Volume:
    0,5 - 10 µl
  • Seq:
    04
  • Tip style:
    Eppendorf type, crystal tip, bevelled
  • Tip colour:
    Transparent
  • For use with:
    Gilson, Eppendorf, Socorex, Nichiryo