Pipette tips
Eppendorf type, with collar, 100 - 1000 µl, -, Transparent, Eppendorf, Nichiryo, Sartorius, Brand, Gilson, Finnpipette, Jencons, Bulk (10×1000)
: DELTALAB
- Packed:Bulk (10×1000)
- Sterile:-
- Volume:100 - 1000 µl
- Seq:19
- Tip style:Eppendorf type, with collar
- Tip colour:Transparent
- For use with:Eppendorf, Nichiryo, Sartorius, Brand, Gilson, Finnpipette, Jencons