VERSA 110 PCR Set-up Workstation Including: VD110 - Deck: 8-position deck with tip priming, waste dispensing and tip dispenser AS100 - Features one robotic arm for liquid handling LD1M-1CDSPTC - Single Channel Dual Syringe Pipettor with Tip Changer - 1 * 1 ST
Catalog # IBSAAU-VER-110-PCR
Supplier: IBS Stock Items
Frequently Bought Together