ContactUsLinkComponent
ProXima 2500 Imaging System Specifications: bit depth: 14bit CCD cooling: no Exposure time: 0.001 - 10s Resolution: 1.8 M pixels Electronic contol (iris, focus, zoom): yes Zoom: 3x Image positioning system: yes Autofocus: yes Automatic door lock: no 1 * 1 ST
Catalog # IBSAIM-520-2500
Supplier:  IBS Stock Items
ProXima 2500 Imaging System Specifications: bit depth: 14bit CCD cooling: no Exposure time: 0.001 - 10s Resolution: 1.8 M pixels Electronic contol (iris, focus, zoom): yes Zoom: 3x Image positioning system: yes Autofocus: yes Automatic door lock: no 1 * 1 ST
Catalog # IBSAIM-520-2500
Supplier:  IBS Stock Items
Frequently Bought Together