ContactUsLinkComponent
Solvent, still head, capacity 1l, height 270mm, cone size 29/32, apparatus for distillation and collection of oxygen free solvents, top stopcock by syringe, drainage through T-bore stockcock, drainage arm utilises a 14/23 Quickfit cone. 1 * 1 items
Catalog # 201-0759
Supplier:  SciLabware
undefined
Solvent, still head, capacity 1l, height 270mm, cone size 29/32, apparatus for distillation and collection of oxygen free solvents, top stopcock by syringe, drainage through T-bore stockcock, drainage arm utilises a 14/23 Quickfit cone. 1 * 1 items
Catalog # 201-0759
Supplier:  SciLabware
Frequently Bought Together