ContactUsLinkComponent
Petri plate, round, height 16.2mm, diameter 90mm, material polystyrene natural, with 1 vent, sterile SAL 10-3. 1 * 700 items
  391-0458
undefined
Petri plate, round, height 16.2mm, diameter 90mm, material polystyrene natural, with 1 vent, sterile SAL 10-3. 1 * 700 items
  391-0458
Frequently Bought Together