ContactUsLinkComponent
Cylinder graduated, capacity 2000ml,hexagonal base with pourout, diameter 95mm,height 420mm, low form, white graduation, class b, graduation divisions 50.0mm. 1 * 6 items
Catalog # KAVA432711746
Supplier:  KAVALIER GLASS
Cylinder graduated, capacity 2000ml,hexagonal base with pourout, diameter 95mm,height 420mm, low form, white graduation, class b, graduation divisions 50.0mm. 1 * 6 items
Catalog # KAVA432711746
Supplier:  KAVALIER GLASS
Frequently Bought Together