ContactUsLinkComponent
Free from toxic heavy metals in formulation. Manufactured to meet ASTM protocols. Tested, certified & batch traceable Density Standa rd 1.0265g/ml @ 25C 1 * 100 mL
  REGEDEN25150
 :  REAGECON
Free from toxic heavy metals in formulation. Manufactured to meet ASTM protocols. Tested, certified & batch traceable Density Standa rd 1.0265g/ml @ 25C 1 * 100 mL
  REGEDEN25150
 :  REAGECON
Frequently Bought Together