ContactUsLinkComponent
Free from toxic heavy metals in formulation. Manufactured to meet ASTM protocols. Tested, certified & batch traceable Density Standa rd 0.8622g/ml @ 25C 1 * 100 mL
Catalog # REGEDEN25110
Supplier:  REAGECON
Free from toxic heavy metals in formulation. Manufactured to meet ASTM protocols. Tested, certified & batch traceable Density Standa rd 0.8622g/ml @ 25C 1 * 100 mL
Catalog # REGEDEN25110
Supplier:  REAGECON
Frequently Bought Together