ContactUsLinkComponent
Free from toxic heavy metals in formulation. Manufactured to meet ASTM protocols. Tested, certified & batch traceable Density Standa rd 2.0812g/ml @ 20C 1 * 100 ml
Catalog # REGEDEN20240
Supplier:  REAGECON
Free from toxic heavy metals in formulation. Manufactured to meet ASTM protocols. Tested, certified & batch traceable Density Standa rd 2.0812g/ml @ 20C 1 * 100 ml
Catalog # REGEDEN20240
Supplier:  REAGECON
Frequently Bought Together