ContactUsLinkComponent
Free from toxic heavy metals in formulation. Manufactured to meet ASTM protocols. Tested, certified & batch traceable Density Standa rd 1.9171g/ml @ 15C 1 * 100 ml
  REGEDEN15230
 :  REAGECON
Free from toxic heavy metals in formulation. Manufactured to meet ASTM protocols. Tested, certified & batch traceable Density Standa rd 1.9171g/ml @ 15C 1 * 100 ml
  REGEDEN15230
 :  REAGECON
Frequently Bought Together