ContactUsLinkComponent
Free from toxic heavy metals in formulation. Manufactured to meet ASTM protocols. Tested, certified & batch traceable Density Standa rd 0.6960g/ml @ 15C 1 * 100 mL
Catalog # REGEDEN15010
Supplier:  REAGECON
Free from toxic heavy metals in formulation. Manufactured to meet ASTM protocols. Tested, certified & batch traceable Density Standa rd 0.6960g/ml @ 15C 1 * 100 mL
Catalog # REGEDEN15010
Supplier:  REAGECON
Frequently Bought Together