0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Slovenia
ContactUsLinkComponent
Sucrose (Brix) Standard 45% - 1.409777 (± 0.00004), Stabilised, shelf-life of 1 year 1 * 15 mL
Catalog # REGEBS45S
Supplier:
REAGECON
Expand image
Sucrose (Brix) Standard 45% - 1.409777 (± 0.00004), Stabilised, shelf-life of 1 year 1 * 15 mL
Catalog # REGEBS45S
Supplier:
REAGECON
Frequently Bought Together
pH buffers, ready to use
Supplier: WTW
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
Sodium hydroxide 30% (w/w) 40% (w/v) in aqueous solution, GPR RECTAPUR® low in nitrogen
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
2,6-Dichlorophenolindophenol sodium salt hydrate ACS analytical reagent
Supplier: VWR Chemicals
Sucrose (Brix) Standard 0% - 1.332986 (± 0.00004), Stabilised, shelf-life of 1 year 1 * 15 mL
Supplier: REAGECON