Sucrose (Brix) Standard 0% - 1.332986 (± 0.00004), Stabilised, shelf-life of 1 year 1 * 15 mL
Catalog # REGEBS00S
Supplier: REAGECON
Sucrose (Brix) Standard 0% - 1.332986 (± 0.00004), Stabilised, shelf-life of 1 year 1 * 15 mL
Catalog # REGEBS00S
Supplier: REAGECON
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR® VP 86, Mini Diaphragm Vacuum Pumps and Compressors
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
2,6-Dichlorophenolindophenol sodium salt hydrate ACS analytical reagent
Supplier: VWR Chemicals
Gallic acid monohydrate ACS
Supplier: Thermo Fisher Scientific
Digital pocket refractometers, PAL series
Supplier: Atago