ContactUsLinkComponent
Sucrose Standard 55% - 1.4308 ( 0.00004) manufactured in accordance with ICUMSA guidelines, tested and certified to 2 decimals plac e. Extended shelf-life of 12 weeks 1 * 15 ml
Catalog # REGEBS55
Supplier:  REAGECON
Sucrose Standard 55% - 1.4308 ( 0.00004) manufactured in accordance with ICUMSA guidelines, tested and certified to 2 decimals plac e. Extended shelf-life of 12 weeks 1 * 15 ml
Catalog # REGEBS55
Supplier:  REAGECON
Frequently Bought Together