ContactUsLinkComponent
Sucrose Standard 45% - 1.409777 ( 0.00004) manufactured in accordance with ICUMSA guidelines, tested and certified to 2 decimals pl ace. Extended shelf-life of 12 weeks 1 * 15 ml
Catalog # REGEBS45
Supplier:  REAGECON
Sucrose Standard 45% - 1.409777 ( 0.00004) manufactured in accordance with ICUMSA guidelines, tested and certified to 2 decimals pl ace. Extended shelf-life of 12 weeks 1 * 15 ml
Catalog # REGEBS45
Supplier:  REAGECON
Frequently Bought Together