ContactUsLinkComponent
Sucrose Standard 12% - 1.35093 ( 0.00004) manufactured in accordance with ICUMSA guidelines, tested and certified to 2 decimals pla ce. Extended shelf-life of 12 weeks 1 * 15 ml
Catalog # REGEBS12
Supplier:  REAGECON
Sucrose Standard 12% - 1.35093 ( 0.00004) manufactured in accordance with ICUMSA guidelines, tested and certified to 2 decimals pla ce. Extended shelf-life of 12 weeks 1 * 15 ml
Catalog # REGEBS12
Supplier:  REAGECON
Frequently Bought Together