ContactUsLinkComponent
Sucrose Standard 7% 1.343253 ( 0.00004) manufactured in accordance with ICUMSA guidelines, tested and certified to 2 decimals place . Extended shelf-life of 12 weeks 1 * 15 ml
Catalog # REGEBS07
Supplier:  REAGECON
Sucrose Standard 7% 1.343253 ( 0.00004) manufactured in accordance with ICUMSA guidelines, tested and certified to 2 decimals place . Extended shelf-life of 12 weeks 1 * 15 ml
Catalog # REGEBS07
Supplier:  REAGECON
Frequently Bought Together