Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Potassium Phosphate pH 6.8 - 961.5ml of Conc. Dilutes 25L 1 * 6 L
Catalog # REGEDBC03-960
Supplier:
REAGECON
Expand image
Potassium Phosphate pH 6.8 - 961.5ml of Conc. Dilutes 25L 1 * 6 L
Catalog # REGEDBC03-960
Supplier:
REAGECON
Frequently Bought Together
Dibutyl ether, extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Acetonitrile 99.9%, ECD tested for residue analysis
Supplier: Thermo Fisher Scientific
2,2,4-Trimethylpentane 99.5% for analysis
Supplier: Thermo Fisher Scientific