Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Simulated Intestinal Fluid, without Enzyme, Ready to use Dissolution Media acc USP & EP 1 * 12 L
Catalog # REGEDB13-121
Supplier:
REAGECON
Expand image
Simulated Intestinal Fluid, without Enzyme, Ready to use Dissolution Media acc USP & EP 1 * 12 L
Catalog # REGEDB13-121
Supplier:
REAGECON
Frequently Bought Together
VWR®, Disposable Weighing Boats
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Iodine 0.05 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Simulated Gastric Fluid, without enzyme, Ready to use Dissolution Media acc USP & EP 1 * 12 L
Supplier: REAGECON