Simulated Gastric Fluid, without enzyme, Ready to use Dissolution Media acc USP & EP 1 * 12 L
Catalog # REGEDB12-121
Supplier: REAGECON
Simulated Gastric Fluid, without enzyme, Ready to use Dissolution Media acc USP & EP 1 * 12 L
Catalog # REGEDB12-121
Supplier: REAGECON
Frequently Bought Together
Potassium hydroxide 0.5 mol/l (0.5 N) in ethanol, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ammonium carbonate, Ultrapure
Supplier: VWR Chemicals