ContactUsLinkComponent
Potassium Phosphate pH 7.5, Ready to use Dissolution Media acc USP & EP 1 * 12 L
Catalog # REGEDB05-121
Supplier:  REAGECON
Potassium Phosphate pH 7.5, Ready to use Dissolution Media acc USP & EP 1 * 12 L
Catalog # REGEDB05-121
Supplier:  REAGECON
Frequently Bought Together