ContactUsLinkComponent
Extension collars for CoolBox XT Orange. Place between the base and original collar to extend the height of the ice-free workstation and enable the use of taller CoolRack sample tube modules. 1 * 1 items
Catalog # 479-1301
Supplier:  BIOCISION
Extension collars for CoolBox XT Orange. Place between the base and original collar to extend the height of the ice-free workstation and enable the use of taller CoolRack sample tube modules. 1 * 1 items
Catalog # 479-1301
Supplier:  BIOCISION
Frequently Bought Together