ContactUsLinkComponent
Cell freezing container for cryopreservation. Alcohol-free. Controlled-rate -1C/min freeze rate to all vials. Use in -80 C freezer. High viability for yeast, cell lines, stem cells, blood, primary cells. Freeze, transfer and store 12 vials 5 mL, wit 1 * 1 items
  479-1182
 :  BIOCISION
undefined
Cell freezing container for cryopreservation. Alcohol-free. Controlled-rate -1C/min freeze rate to all vials. Use in -80 C freezer. High viability for yeast, cell lines, stem cells, blood, primary cells. Freeze, transfer and store 12 vials 5 mL, wit 1 * 1 items
  479-1182
 :  BIOCISION
Frequently Bought Together