Cell freezing container for cryopreservation. Alcohol-free. Controlled-rate -1C/min freeze rate to all vials. Use in -80 C freezer. High viability for yeast, cell lines, stem cells, blood, primary cells. Freeze, transfer and store 12 vials 1,5 or 2 1 * 1 items
Catalog # 479-1179
Supplier: BIOCISION
Frequently Bought Together