ContactUsLinkComponent
Multi Vortex Mixer with Platform to suit 16 x 1.5mL, 8 x 0.5mL and 8 x 0.2mL Tubes - Each 1 * 1 items
Catalog # 444-1035
Supplier:  GRANT INSTRUMENTS
Multi Vortex Mixer with Platform to suit 16 x 1.5mL, 8 x 0.5mL and 8 x 0.2mL Tubes - Each 1 * 1 items
Catalog # 444-1035
Supplier:  GRANT INSTRUMENTS
Frequently Bought Together