Scoops
90 ml, 180 mm, HDPE, White
Catalog # LAMA62
Supplier: LAMAPLAST
Jump to:
Product Details & Documents
These scoops are made of high density PE. Ideal for use in food applications.
- For withdrawal of solid and granulates samples
- Rounded bottom
: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Documents
TDS Powder Scoops
Datasheet
Specifications
- Capacity:90 ml
- Colour:White
- Length:180 mm
- Material:HDPE
- Seq:01